Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluoro-5-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

33320-1G 33320-25G 33320-500MG 33320-5G
76931-152EA 401.03 USD InStock
76931-152 76931-154 76931-156 76931-158
4-Chloro-2-fluoro-5-methylphenylboronic acid pinacol ester ≥97%
4-Chloro-2-fluoro-5-methylphenylboronic acid pinacol ester


Formula: C₁₃H₁₇BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD11855985
CAS Number: 1126320-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty