Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyclopropylphenylboronic Acid Pinacol Ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

33645-1G 33645-25G 33645-500MG 33645-5G
76931-216EA 188.85 USD InStock
76931-216 76931-218 76931-220 76931-222
4-Cyclopropylphenylboronic Acid Pinacol Ester ≥97% research grade
4-Cyclopropylphenylboronic Acid Pinacol Ester


Formula: C₁₅H₂₁BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD10698529
CAS Number: 1219741-94-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty