Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Hydroxy-[1,1'-biphenyl]-4-carbaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

34691-1G 34691-25G 34691-500MG 34691-5G
76931-446EA 189.91 USD InStock
76931-446 76931-448 76931-450 76931-452
4'-Hydroxy-[1,1'-biphenyl]-4-carbaldehyde ≥97% research grade
4'-Hydroxy-[1,1'-biphenyl]-4-carbaldehyde


Formula: C₁₃H₁₀O₂
MW: 198.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00804656
CAS Number: 100980-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty