Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-3-methoxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

36375-1G 36375-25G 36375-500MG 36375-5G
76931-826EA 184.6 USD InStock
76931-826 76931-828 76931-830 76931-832
4-Cyano-3-methoxybenzoic acid ≥97% research grade
4-Cyano-3-methoxybenzoic acid


Formula: C₉H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD18203531
CAS Number: 102362-00-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty