Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Methoxy-3-methyl-1,1'-biphenyl ≥97%

Supplier: AOB CHEM USA
Warning

36782-1G 36782-25G 36782-500MG 36782-5G
76931-906EA 284.33 USD InStock
76931-906 76931-908 76931-910 76931-912
4'-Methoxy-3-methyl-1,1'-biphenyl ≥97%
4'-Methoxy-3-methyl-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₁₄H₁₄O
Storage Temperature: Refrigerator
MDL Number: MFCD06201372
CAS Number: 17171-17-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty