Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Methoxybiphenyl-4-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

36964-1G 36964-25G 36964-500MG 36964-5G
76931-956EA 292.81 USD InStock
76931-956 76931-958 76931-960 76931-962
4'-Methoxybiphenyl-4-carboxylic acid ≥97%
4'-Methoxybiphenyl-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₃
MW: 228.25
Storage Temperature: Ambient
MDL Number: MFCD00185792
CAS Number: 725-14-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty