Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-4-methoxy-2-methylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

37865-1G 37865-25G 37865-500MG 37865-5G
76932-126EA 439.22 USD InStock
76932-126 76932-128 76932-130 76932-132
5-Fluoro-4-methoxy-2-methylbenzaldehyde ≥97%
5-Fluoro-4-methoxy-2-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD22414669
CAS Number: 1781014-55-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty