Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-4-methoxy-2-methylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

39302-1G 39302-25G 39302-500MG 39302-5G
76932-310EA 287.51 USD InStock
76932-310 76932-312 76932-314 76932-316
5-Chloro-4-methoxy-2-methylphenylboronic acid ≥97% research grade
5-Chloro-4-methoxy-2-methylphenylboronic acid


Formula: C₈H₁₀BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09054938
CAS Number: 511295-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty