Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-isopropoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

39802-1G 39802-25G 39802-500MG 39802-5G
76932-326EA 124.13 USD InStock
76932-326 76932-328 76932-330 76932-332
4-Chloro-2-isopropoxyphenylboronic acid ≥97%
4-Chloro-2-isopropoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₂BClO₃
Storage Temperature: Ambient
MDL Number: MFCD17015770
CAS Number: 1256355-04-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty