Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-1-benzothiophene-2-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

45730-1G 45730-25G 45730-500MG 45730-5G
76932-524EA 192.03 USD InStock
76932-524 76932-526 76932-528 76932-530
5-Chloro-1-benzothiophene-2-carboxylic acid ≥97%
5-Chloro-1-benzothiophene-2-carboxylic acid


Formula: C₉H₅ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD01927183
CAS Number: 13771-75-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty