Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-2-isopropylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

46901-1G 46901-25G 46901-500MG 46901-5G
76932-690EA 423.3 USD InStock
76932-690 76932-692 76932-694 76932-696
4-Hydroxy-2-isopropylbenzaldehyde ≥97%
4-Hydroxy-2-isopropylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16997130
CAS Number: 181035-58-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty