Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxy-2,6-dimethylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

48997-1G 48997-25G 48997-500MG 48997-5G
76932-864EA 350.1 USD InStock
76932-864 76932-866 76932-868 76932-870
4-Ethoxy-2,6-dimethylbenzoic acid ≥97%
4-Ethoxy-2,6-dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD14537280
CAS Number: 91970-50-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty