Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(tert-Butyl)-2-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

52128-1G 52128-25G 52128-500MG 52128-5G
76933-094EA 411.63 USD InStock
76933-094 76933-096 76933-098 76933-100
5-(tert-Butyl)-2-methylbenzoic acid ≥97%
5-(tert-Butyl)-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₂
MW: 192.254 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09260803
CAS Number: 28162-25-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty