Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(3-Chloro-5-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOB CHEM USA
Warning

55264-1G 55264-25G 55264-500MG 55264-5G
76933-700EA 375.56 USD InStock
76933-700 76933-702 76933-704 76933-706
5-(3-Chloro-5-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Chloro-5-fluorophenyl)thiazol-2-amine
Precaución: for Research Use Only


Formula: C₉H₆ClFN₂S
MW: 228.674 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24221416
CAS Number: 1552432-91-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty