Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-5-formyl-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

56332-1G 56332-500MG 56332-5G
76934-188EA 1089.55 USD InStock
76934-188 76934-190 76934-192
4-Fluoro-5-formyl-2-(trifluoromethyl)benzoic acid ≥97%
4-Fluoro-5-formyl-2-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₄F₄O₃
MW: 236.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564355
CAS Number: 2387341-88-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty