Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formyl-1-methyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

60151-1G 60151-25G 60151-500MG 60151-5G
76935-064EA 344.8 USD InStock
76935-064 76935-066 76935-068 76935-070
5-Formyl-1-methyl-1H-pyrazole-4-carboxylic acid ≥97%
5-Formyl-1-methyl-1H-pyrazole-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₆N₂O₃
MW: 154.123 g/mol
Storage Temperature: Ambient
CAS Number: 1368337-66-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty