Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2,3-dimethylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

64425-1G 64425-25G 64425-500MG 64425-5G
76935-610EA 338.43 USD InStock
76935-610 76935-612 76935-614 76935-616
4-Fluoro-2,3-dimethylbenzoic acid ≥97% research grade
4-Fluoro-2,3-dimethylbenzoic acid


Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD23712108
CAS Number: 1104380-03-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty