Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,3-difluoroisonicotinaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

64696-1G 64696-25G 64696-500MG 64696-5G
76935-636EA 277.96 USD InStock
76935-636 76935-638 76935-640 76935-642
5-Chloro-2,3-difluoroisonicotinaldehyde ≥97%
5-Chloro-2,3-difluoroisonicotinaldehyde
Precaución: for Research Use Only


Formula: C₆H₂ClF₂NO
Storage Temperature: Refrigerator
CAS Number: 1333319-52-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty