Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-chloro-2-isopropoxybenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

64790-1G 64790-25G 64790-500MG 64790-5G
76935-652EA 375.56 USD InStock
76935-652 76935-654 76935-656 76935-658
5-Bromo-3-chloro-2-isopropoxybenzaldehyde ≥97%
5-Bromo-3-chloro-2-isopropoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₀BrClO₂
MW: 277.5422 g/mol
Storage Temperature: Refrigerator
CAS Number: 2469839-22-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty