Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-fluoro-1-naphthoate ≥97%

Supplier: AOB CHEM USA
Warning

12268-1G 12268-25G 12268-500MG 12268-5G
76936-514EA 352.22 USD InStock
76936-514 76936-516 76936-518 76936-520
Ethyl 4-fluoro-1-naphthoate ≥97%
Ethyl 4-fluoro-1-naphthoate
Precaución: for Research Use Only


Formula: C₁₃H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11555051
CAS Number: 1512-51-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty