Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-4-butoxyphenyltrifluoroborate ≥97%

Supplier: AOB CHEM USA
Warning

12403-25G 12403-5G
76936-552EA 456.19 USD InStock
76936-552 76936-554
Potassium-4-butoxyphenyltrifluoroborate ≥97%
Potassium 4-butoxyphenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₁₀H₁₃BF₃KO
Storage Temperature: Ambient
MDL Number: MFCD04115755
CAS Number: 850623-61-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty