Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoropyridin-3-ol ≥97% research grade

Supplier: AOB CHEM USA
Warning

13553-1G 13553-25G 13553-500MG 13553-5G
76936-802EA 153.84 USD InStock
76936-802 76936-804 76936-806 76936-808
6-Bromo-2-fluoropyridin-3-ol ≥97% research grade
6-Bromo-2-fluoropyridin-3-ol


Formula: C₅H₃BrFNO
MW: 191.986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999156
CAS Number: 850142-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty