Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-amino-7-bromobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

136358-1G 136358-500MG 136358-5G 136358-25G
76936-816EA 1028.02 USD InStock
76936-816 76936-818 76936-820 77653-476
Ethyl 3-amino-7-bromobenzo[b]thiophene-2-carboxylate ≥97%
Ethyl 3-amino-7-bromobenzo[b]thiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₀BrNO₂S
MW: 300.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29141527
CAS Number: 2091306-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty