Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3,4-difluoro-2-methoxyaniline ≥95%

Supplier: AOB CHEM USA
Warning

16559-1G 16559-25G 16559-500MG 16559-5G
76937-138EA 266.29 USD InStock
76937-138 76937-140 76937-142 76937-144
6-Bromo-3,4-difluoro-2-methoxyaniline ≥95%
6-Bromo-3,4-difluoro-2-methoxyaniline
Precaución: for Research Use Only


Formula: C₇H₆BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD7368890
CAS Number: 2573784-23-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty