Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-pyridineboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

16924-1G 16924-25G 16924-5G
76937-160EA 64.72 USD InStock
76937-160 76937-162 76937-164
2-Benzyloxy-5-pyridineboronic acid ≥97%
2-Benzyloxy-5-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₂BNO₃
MW: 229.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06798262
CAS Number: 929250-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty