Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Isopropoxy-5-methylnicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

17999-1G 17999-25G 17999-500MG 17999-5G
76937-254EA 168.69 USD InStock
76937-254 76937-256 76937-258 76937-260
6-Isopropoxy-5-methylnicotinic acid ≥97%
6-Isopropoxy-5-methylnicotinic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13181628
CAS Number: 1011558-18-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty