Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Ethoxy-2-methylpyridine-3-boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

21278-1G 21278-25G 21278-500MG 21278-5G
76937-818EA 290.69 USD InStock
76937-818 76937-820 76937-822 76937-824
6-Ethoxy-2-methylpyridine-3-boronic acid ≥97%
6-Ethoxy-2-methylpyridine-3-boronic acid
Precaución: for Research Use Only


Formula: C₈H₁₂BNO₃
MW: 180.997 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977794
CAS Number: 1451391-75-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty