Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-2-methylnicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

21299-1G 21299-25G 21299-500MG 21299-5G
76937-830EA 159.14 USD InStock
76937-830 76937-832 76937-834 76937-836
6-Methoxy-2-methylnicotinic acid ≥97% research grade
6-Methoxy-2-methylnicotinic acid


Formula: C₈H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD16611508
CAS Number: 1227515-71-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty