Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzenemethanol, 3-bromo-2-hydroxy-5-methyl- ≥97%

Supplier: AOB CHEM USA
Warning

21755-1G 21755-25G 21755-500MG 21755-5G
76937-988EA 147.47 USD InStock
76937-988 76937-990 76937-992 76937-994
Benzenemethanol, 3-bromo-2-hydroxy-5-methyl- ≥97%
Benzenemethanol, 3-bromo-2-hydroxy-5-methyl-
Precaución: for Research Use Only


Formula: C₈H₉BrO₂
Storage Temperature: Ambient
CAS Number: 43135-49-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty