Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-2-(trifluoromethoxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

22552-1G 22552-25G 22552-500MG 22552-5G
76938-124EA 201.58 USD InStock
76938-124 76938-126 76938-128 76938-130
Ethyl 5-bromo-2-(trifluoromethoxy)benzoate ≥97%
Ethyl 5-bromo-2-(trifluoromethoxy)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₈BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD06204542
CAS Number: 773135-66-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty