Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloroisonicotinic acid ethyl ester ≥95% research grade

Supplier: AOB CHEM USA
Warning

25991-25G 25991-5G
76938-742EA 289.63 USD InStock
76938-742 76938-744
2,6-Dichloroisonicotinic acid ethyl ester ≥95% research grade
2,6-Dichloroisonicotinic acid ethyl ester


Formula: C₈H₇Cl₂NO₂
MW: 220.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235122
CAS Number: 1604-14-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty