Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylsulfanylpyridine-3-Carboxylic Acid ≥97%

Supplier: AOB CHEM USA
Warning

27292-1G 27292-25G 27292-500MG 27292-5G
76939-074EA 141.1 USD InStock
76939-074 76939-076 76939-078 76939-080
6-Methylsulfanylpyridine-3-Carboxylic Acid ≥97%
6-Methylsulfanylpyridine-3-Carboxylic Acid
Precaución: for Research Use Only


Formula: C₇H₇NO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD01860214
CAS Number: 74470-25-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty