Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-8-iodoimidazo[1,2-a]pyridine 97 Research

Supplier: AOB CHEM USA
Warning

27817-1G 27817-25G 27817-5G
76939-220EA 91.24 USD InStock
76939-220 76939-222 76939-224
6-Bromo-8-iodoimidazo[1,2-a]pyridine 97 Research
6-Bromo-8-iodoimidazo[1,2-a]pyridine


Formula: C₇H₄BrIN₂
MW: 322.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21609484
CAS Number: 1364917-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty