Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(5-Bromopyridin-2-yl)benzamide ≥97%

Supplier: AOB CHEM USA
Warning

28071-1G 28071-25G 28071-500MG 28071-5G
76939-264EA 175.05 USD InStock
76939-264 76939-266 76939-268 76939-270
N-(5-Bromopyridin-2-yl)benzamide ≥97%
N-(5-Bromopyridin-2-yl)benzamide
Precaución: for Research Use Only


Formula: C₁₂H₉BrN₂O
Storage Temperature: Ambient
CAS Number: 62802-75-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty