Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-bromo-5-fluoroisonicotinate ≥97%

Supplier: AOB CHEM USA
Warning

28706-1G 28706-25G 28706-500MG 28706-5G
76939-362EA 254.62 USD InStock
76939-362 76939-364 76939-366 76939-368
Ethyl 3-bromo-5-fluoroisonicotinate ≥97%
Ethyl 3-bromo-5-fluoroisonicotinate
Precaución: for Research Use Only


Formula: C₈H₇BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD14698327
CAS Number: 1214335-25-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty