Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium quinoline-6-trifluoroborate ≥97%

Supplier: AOB CHEM USA
Warning

28761-1G 28761-25G 28761-500MG 28761-5G
76939-376EA 124.13 USD InStock
76939-376 76939-378 76939-380 76939-382
Potassium quinoline-6-trifluoroborate ≥97%
Potassium quinoline-6-trifluoroborate
Precaución: for Research Use Only


Formula: C₉H₆BF₃KN
Storage Temperature: Refrigerator
MDL Number: MFCD09993442
CAS Number: 1411985-98-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty