Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-5-methoxypyridine-3-carbaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

29390-1G 29390-25G 29390-500MG 29390-5G
76939-456EA 379.81 USD InStock
76939-456 76939-458 76939-460 76939-462
6-Bromo-5-methoxypyridine-3-carbaldehyde ≥97%
6-Bromo-5-methoxypyridine-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD18257273
CAS Number: 1256833-92-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty