Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chloro-4,5-difluorobenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

29472-1G 29472-25G 29472-500MG 29472-5G
76939-466EA 142.17 USD InStock
76939-466 76939-468 76939-470 76939-472
Methyl-2-chloro-4,5-difluorobenzoate ≥97% research grade
Methyl-2-chloro-4,5-difluorobenzoate


Formula: C₈H₅ClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD10566827
CAS Number: 128800-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty