Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-fluoro-3-iodopyridine ≥97%

Supplier: AOB CHEM USA
Warning

29627-1G 29627-25G 29627-500MG 29627-5G
76939-486EA 254.62 USD InStock
76939-486 76939-488 76939-490 76939-492
6-Chloro-2-fluoro-3-iodopyridine ≥97%
6-Chloro-2-fluoro-3-iodopyridine
Precaución: for Research Use Only


Formula: C₅H₂ClFIN
MW: 257.43
Storage Temperature: Refrigerator
MDL Number: MFCD12922962
CAS Number: 1187732-65-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty