Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-4-methylpyridin-3-ol ≥95%

Supplier: AOB CHEM USA
Warning

30890-1G 30890-25G 30890-500MG 30890-5G
76939-788EA 274.78 USD InStock
76939-788 76939-790 76939-792 76939-794
6-Bromo-4-methylpyridin-3-ol ≥95%
6-Bromo-4-methylpyridin-3-ol
Precaución: for Research Use Only


Formula: C₆H₆BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD18255424
CAS Number: 1256824-49-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty