Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-methoxy-4-methylnicotinate ≥97%

Supplier: AOB CHEM USA
Warning

31289-1G 31289-500MG 31289-5G
76939-892EA 647.15 USD InStock
76939-892 76939-894 76939-896
Methyl 5-methoxy-4-methylnicotinate ≥97%
Methyl 5-methoxy-4-methylnicotinate
Precaución: for Research Use Only


Formula: C₉H₁₁NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18910327
CAS Number: 1378678-88-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty