Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-(4-Bromo-2-Nitro-Phenyl)-Sulfide ≥97%

Supplier: AOB CHEM USA
Warning

31779-1G 31779-25G 31779-500MG 31779-5G
76940-012EA 144.29 USD InStock
76940-012 76940-014 76940-016 76940-018
Ethyl-(4-Bromo-2-Nitro-Phenyl)-Sulfide ≥97%
Ethyl-(4-Bromo-2-Nitro-Phenyl)-Sulfide
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂S
MW: 262.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22488427
CAS Number: 900175-18-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty