Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Butoxy-5-methylpyridine-3-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

33420-1G 33420-25G 33420-500MG 33420-5G
76940-454EA 350.1 USD InStock
76940-454 76940-456 76940-458 76940-460
6-Butoxy-5-methylpyridine-3-carboxylic acid ≥97%
6-Butoxy-5-methylpyridine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD30834059
CAS Number: 2027444-93-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty