Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-fluoro-4-(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

33504-1G 33504-25G 33504-500MG 33504-5G
76940-484EA 147.47 USD InStock
76940-484 76940-486 76940-488 76940-490
Ethyl 2-fluoro-4-(trifluoromethyl)benzoate ≥97%
Ethyl 2-fluoro-4-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₈F₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06204411
CAS Number: 702673-00-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty