Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-hydroxy-3-isopropylbenzoate ≥95% research grade

Supplier: AOB CHEM USA
Warning

34275-1G 34275-25G 34275-500MG 34275-5G
76940-714EA 212.18 USD InStock
76940-714 76940-716 76940-718 76940-720
Methyl 2-hydroxy-3-isopropylbenzoate ≥95% research grade
Methyl 2-hydroxy-3-isopropylbenzoate


Formula: C₁₁H₁₄O₃
MW: 194.2271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24714020
CAS Number: 52159-64-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty