Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoro-3-hydroxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

34407-1G 34407-25G 34407-500MG 34407-5G
76940-788EA 374.5 USD InStock
76940-788 76940-790 76940-792 76940-794
6-Bromo-2-fluoro-3-hydroxybenzoic acid ≥97%
6-Bromo-2-fluoro-3-hydroxybenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄BrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28738982
CAS Number: 91659-33-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty