Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-bromo-5-fluoro-2-methylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

34495-1G 34495-25G 34495-500MG 34495-5G
76940-802EA 194.15 USD InStock
76940-802 76940-804 76940-806 76940-808
Ethyl 4-bromo-5-fluoro-2-methylbenzoate ≥97%
Ethyl 4-bromo-5-fluoro-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28739714
CAS Number: 1807043-81-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty