Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-(piperidin-1-yl)nicotinate ≥97%

Supplier: AOB CHEM USA
Warning

34571-1G 34571-25G 34571-500MG 34571-5G
76940-818EA 132.62 USD InStock
76940-818 76940-820 76940-822 76940-824
Methyl 6-(piperidin-1-yl)nicotinate ≥97%
Methyl 6-(piperidin-1-yl)nicotinate
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD05664432
CAS Number: 132521-82-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty