Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,6-dichloro-4-(trifluoromethoxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

34580-1G 34580-25G 34580-500MG 34580-5G
76940-834EA 225.98 USD InStock
76940-834 76940-836 76940-838 76940-840
Methyl 2,6-dichloro-4-(trifluoromethoxy)benzoate ≥97%
Methyl 2,6-dichloro-4-(trifluoromethoxy)benzoate
Precaución: for Research Use Only


Formula: C₉H₅Cl₂F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD31619504
CAS Number: 2221812-05-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty