Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-chloro-2-methoxypyridine-4-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

34649-1G 34649-25G 34649-5G
76940-920EA 107.16 USD InStock
76940-920 76940-922 76940-924
Ethyl 5-chloro-2-methoxypyridine-4-carboxylate ≥97%
Ethyl 5-chloro-2-methoxypyridine-4-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₀ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD14698196
CAS Number: 1214345-87-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty